missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alcohol dehydrogenase 1B Protein
A DNA sequence encoding the Homo sapiens (Human) Alcohol dehydrogenase 1B, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_000659.2 |
---|---|
Gene ID (Entrez) | 125 |
Name | Alcohol dehydrogenase 1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936439
|
enQuireBio™
QP5613-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956439
|
enQuireBio™
QP5613-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946439
|
enQuireBio™
QP5613-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15966439
|
enQuireBio™
QP5613-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15976439
|
enQuireBio™
QP5613-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15986439
|
enQuireBio™
QP5613-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000659.2 | |
Alcohol dehydrogenase 1B Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
125 | |
Research Use Only | |
ADH1B | |
Recombinant Protein | |
E. coli | |
STAGKVIKCK AAVLWEVKKP FSIEDVEVAP PKAYEVRIKM VAVGICRTDD HVVSGNLVTP LPVILGHEAA GIVESVGEGV TTVKPGDKVI PLFTPQCGKC RVCKNPESNY CLKNDLGNPR GTLQDGTRRF TCRGKPIHHF LGTSTFSQYT VVDENAVAKI DAASPLEKVC LIGCGFSTGY GSAVNVAKVT PGSTCAVFGL GGVGLSAVMG CKAAGAARII AVDINKDKFA KAKELGATEC INPQDYKKPI QEVLKEMTDG GVDFSFEVIG RLDTMMASLL CCHEACGTSV IVGVPPASQN LSINPMLLLT GRTWKGAVYG GFKSKEGIPK LVADFMAKKF SLDALITHVL PFEKINEGFD LLHSGKSIRT VLTF | |
Greater than 90% as determined by SDS-PAGE. |