missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Alpha-tocopherol transfer Protein

Product Code. p-7153696
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16016260 10 μg 10µg
16056260 50 μg 50µg
16006260 100 μg 100µg
16036260 200 μg 200µg
16046260 500 μg 500µg
16026260 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16016260 Supplier enQuireBio™ Supplier No. QP6847ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Alpha-tocopherol transfer protein, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000361.1
Gene ID (Entrez) 7274
Name Alpha-tocopherol transfer Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TTPA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MAEARSQPSA GPQLNALPDH SPLLQPGLAA LRRRAREAGV PLAPLPLTDS FLLRFLRARD FDLDLAWRLL KNYYKWRAEC PEISADLHPR SIIGLLKAGY HGVLRSRDPT GSKVLIYRIA HWDPKVFTAY DVFRVSLITS ELIVQEVETQ RNGIKAIFDL EGWQFSHAFQ ITPSVAKKIA AVLTDSFPLK VRGIHLINEP VIFHAVFSMI KPFLTEKIKE RIHMHGNNYK QSLLQHFPDI LPLEYGGEEF SMEDICQEWT NFIMKSEDYL SSISESIQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.