missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Amino-terminal enhancer of split Protein

A DNA sequence encoding the Homo sapiens (Human) Amino-terminal enhancer of split, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5618-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956479

  • 505.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001121.2
Amino-terminal enhancer of split Protein
Research Use Only
AES
Recombinant Protein
E. coli
MCHKNGFPQE GGITAAFLQK RKLRLSKNHR PARAKVTEHV RGTRPGRATA GPAASTRAAG SLFFDRWGNR GPAGCRGSSH LPQQLKFTTS DSCDRIKDEF QLLQAQYHSL KLECDKLASE KSEMQRHYVM YYEMSYGLNI EMHKQAEIVK RLNGICAQVL PYLSQEHQQQ VLGAIERAKQ VTAPELNSII RQQLQAHQLS QLQALALPLT PLPVGLQPPS LPAVSAGTGL LSLSALGSQA HLSKEDKNGH DGDTHQEDDG EKSD
Greater than 80% as determined by SDS-PAGE.
166
100 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers