missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Anaphase-promoting complex subunit 15 Protein
A DNA sequence encoding the Homo sapiens (Human) Anaphase-promoting complex subunit 15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5732-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265414.1 | |
Anaphase-promoting complex subunit 15 Protein | |
Research Use Only | |
ANAPC15 | |
Recombinant Protein | |
E. coli | |
MSTLFPSLFP RVTETLWFNL DRPCVEETEL QQQEQQHQAW LQSIAEKDNN LVPIGKPASE HYDDEEEEDD EDDEDSEEDS EDDEDMQDMD EMNDYNESPD DGEVNEVDME GNEQDQDQWM I | |
Greater than 90% as determined by SDS-PAGE. |
25906 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |