missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ang K1-3 Protein

Product Code. 15904128
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15904128 20 μg 20µg
15984118 100 μg 100µg
15994118 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15904128 Supplier enQuireBio™ Supplier No. QP1052020ug
  • ONLINE EXCLUSIVE PRICE
    150.15 €
    140.98 € / 20µg
    Save 9.17 €
    6% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cDNA sequence encoding the Ang K1-3 was constructed and used to recombinantly synthesize the protein.

Specifications

Name Ang K1-3 Protein
Quantity 20 μg
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The activity is assayed on anti-proliferation and anti-migration of endothelial cells in vitro and anti-angiogenesis in vivo. The specific activity of anti-migration of endothelial cells in vitro is 55,000 Units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence VYLSECKTGN GKNYRGTMSK TKNGITCQKW SSTSPHRPRF SPATHPSEGL EENYCRNPDN DPQGPWCYTT DPEKRYDYCD ILECEEECMH CSGENYDGKI SKTMSGLECQ AWDSQSPHAH GYIPSKFPNK NLKKNYCRNP DRELRPWCFT TDPNKRWELC DIPRCTTPPP SSGPTYQCLKGTGENYRGNV AVTVSGHTCQ HWSAQTPHTH NRTPENFPCK NLDENYCRNP DGKRAPWCHT TNSQVRWEYC KIPSCDSSP
Buffer Lyophilized from a 0.2?m filtered concentrated (1.0 mg/ml) solution in 20mM NaAc, pH5.5, 4% mannitol.
Purity or Quality Grade Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.