missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOA1 Protein
A cDNA sequence encoding the APOA1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 870.00 €
Specifications
Gene ID (Entrez) | 335 |
---|---|
Name | APOA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | APOA1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995958
|
enQuireBio™
QP11043-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975958
|
enQuireBio™
QP11043-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985958
|
enQuireBio™
QP11043-1MG |
1 mg |
870.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
335 | |
Research Use Only | |
APOA1 | |
Human | |
Untagged | |
The APOA1 1 mg/ml solution contains 10mM Ammonium Bicarbonate at pH 7.4. |
APOA1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MKAAVLTLAV LFLTGSQARH FWQQDEPPQS PWDRVKDLAT VYVDVLKDSG RDYVSQFEGS ALGKQLNLKL LDNWDSVTST FSKLREQLGP VTQEFWDNLE KETEGLRQEM SKDLEEVKAK VQPYLDDFQK KWQEEMELYR QKVEPLRAEL QEGARQKLHE LQEKLSPLGE EMRDRARAHV DALRTHLAPY SDELRQRLAA RLEALKENGG ARLAEYHAKA TEHLSTLSEK AKPALEDLRQ GLLPVLESFK VSFLSALEEY TKKLNTQ | |
Greater than 95.0% as determined by SDS-PAGE. |