missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human APOA5 Protein

Product Code. 15925978
Click to view available options
Quantity:
10 μg
2 μg
Unit Size:
10µg
2µg
This item is not returnable. View return policy

Product Code. 15925978

Brand: enQuireBio™ QP11046HIS2ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the APOA5 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 116519
Name APOA5 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol APOA5
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence RKGFWDYFSQ TSGDKGRVEQ IHQQKMAREP ATLKDSLEQD LNNMNKFLEK LRPLSGSEAP RLPQDPVGMR RQLQEELEEV KARLQPYMAE AHELVGWNLE GLRQQLKPYT MDLMEQVALR VQELQEQLRV VGEDTKAQLL GGVDEAWALL QGLQSRVVHH TGRFKELFHP YAESLVSGIG RHVQELHRSV APHAPASPAR LSRCVQVLSR KLTLKAKALH ARIQQNLDQL REELSRAFAG TGTEEGAGPD PQMLSEEVRQ RLQAFRQDTY LQIAAFTRAI DQETEEVQQQ LAPPPPGHSA FAPEFQQTDS GKVLSKLQAR LDDLWEDITH SLHDQGHSHL GDPAAADYKD DDDK
Buffer APOA5 Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS and 50mM NaCl, pH 7.5
Purity or Quality Grade Greater than 70% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.