missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human APOBEC4 Protein

A cDNA sequence encoding the APOBEC4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11048-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965978

  • 3915.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

APOBEC4 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEPIYEE YLANHGTIVK PYYWLSFSLD CSNCPYHIRT GEEARVSLTE FCQIFGFPYG TTFPQTKHLT FYELKTSSGS LVQKGHASSC TGNYIHPESM LFEMNGYLDS AIYNNDSIRH IILYSNNSPC NEANHCCISK MYNFLITYPG ITLSIYFSQL YHTEMDFPAS AWNREALRSL ASLWPRVVLS PISGGIWHSV LHSFISGVSG SHVFQPILTG RALADRHNAY EINAITGVKP YFTDVLLQTK RNPNTKAQEA LESYPLNNAF PGQFFQMPSG QLQPNLPPDL RAPVVFVLVP LRDLPPMHMG QNPNKPRNIV RHLNMPQMSF QETKDLGRLP TGRSVEIVEI TEQFASSKEA DEKKKKKGKK
Greater than 85% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
APOBEC4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers