missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOBEC4 Protein
A cDNA sequence encoding the APOBEC4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11048-20ug
Additional Details : Weight : 0.01000kg
Specifications
APOBEC4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPIYEE YLANHGTIVK PYYWLSFSLD CSNCPYHIRT GEEARVSLTE FCQIFGFPYG TTFPQTKHLT FYELKTSSGS LVQKGHASSC TGNYIHPESM LFEMNGYLDS AIYNNDSIRH IILYSNNSPC NEANHCCISK MYNFLITYPG ITLSIYFSQL YHTEMDFPAS AWNREALRSL ASLWPRVVLS PISGGIWHSV LHSFISGVSG SHVFQPILTG RALADRHNAY EINAITGVKP YFTDVLLQTK RNPNTKAQEA LESYPLNNAF PGQFFQMPSG QLQPNLPPDL RAPVVFVLVP LRDLPPMHMG QNPNKPRNIV RHLNMPQMSF QETKDLGRLP TGRSVEIVEI TEQFASSKEA DEKKKKKGKK | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
APOBEC4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |