missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human APOE3 Protein

Product Code. 15954128
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 15954128

Brand: enQuireBio™ QP105221mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the APOE3 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 348
Name APOE3 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol APOE
Biological Activity Immobilized rHuApoE3 binds to rMuVLDLR with EC50 less than 0.4microg/ml.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MHHHHHHKVE QAVETEPEPE LRQQTEWQSG QRWELALGRF WDYLRWVQTL SEQVQEELLS SQVTQELRAL MDETMKELKA YKSELEEQLT PVAEETRARL SKELQAAQAR LGADMEDVCG RLVQYRGEVQ AMLGQSTEEL RVRLASHLRK LRKRLLRDAD DLQKRLAVYQ AGAREGAERG LSAIRERLGP LVEQGRVRAA TVGSLAGQPL QERAQAWGER LRARMEEMGS RTRDRLDEVK EQVAEVRAKL EEQAQQIRLQ AEAFQARLKS WFEPLVEDMQ RQWAGLVEKV QAAVGTSAAP VPSDNH
Buffer Lyophilized from a 0.2microm filtered solution in PBS.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.