missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOE3 Protein
A cDNA sequence encoding the APOE3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10522-500ug
Additional Details : Weight : 0.01000kg
Specifications
348 | |
500 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Immobilized rHuApoE3 binds to rMuVLDLR with EC50 less than 0.4microg/ml. | |
Human | |
His | |
Lyophilized from a 0.2microm filtered solution in PBS. |
APOE3 Protein | |
Research Use Only | |
APOE | |
Recombinant Protein | |
E. coli | |
MHHHHHHKVE QAVETEPEPE LRQQTEWQSG QRWELALGRF WDYLRWVQTL SEQVQEELLS SQVTQELRAL MDETMKELKA YKSELEEQLT PVAEETRARL SKELQAAQAR LGADMEDVCG RLVQYRGEVQ AMLGQSTEEL RVRLASHLRK LRKRLLRDAD DLQKRLAVYQ AGAREGAERG LSAIRERLGP LVEQGRVRAA TVGSLAGQPL QERAQAWGER LRARMEEMGS RTRDRLDEVK EQVAEVRAKL EEQAQQIRLQ AEAFQARLKS WFEPLVEDMQ RQWAGLVEKV QAAVGTSAAP VPSDNH | |
Greater than 95% as determined by SDS-PAGE. |