missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOE4 Protein
A cDNA sequence encoding the APOE4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11051-100ug
Additional Details : Weight : 0.01000kg
Specifications
348 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
APOE | |
Human | |
His | |
Lyophilized from a sterile (0.2microm) filtered aqueous solution containing 10mM sodium phosphate, pH 7.5. |
APOE4 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MHHHHHHKVE QAVETEPEPE LRQQTEWQSG QRWELALGRF WDYLRWVQTL SEQVQEELLS SQVTQELRAL MDETMKELKA YKSELEEQLT PVAEETRARL SKELQAAQAR LGADMEDVRG RLVQYRGEVQ AMLGQSTEEL RVRLASHLRK LRKRLLRDAD DLQKRLAVYQ AGAREGAERG LSAIRERLGP LVEQGRVRAA TVGSLAGQPL QERAQAWGER LRARMEEMGS RTRDRLDEVK EQVAEVRAKL EEQAQQIRLQ AEAFQARLKS WFEPLVEDMQ RQWAGLVEKV QAAVGTSAAP VPSDNH |