missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Arginase-2, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Arginase-2, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5676-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986919

  • 2285.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001163.1
Arginase-2, mitochondrial Protein
Research Use Only
Arg2
Recombinant Protein
E. coli
VHSVAVIGAP FSQGQKRKGV EHGPAAIREA GLMKRLSSLG CHLKDFGDLS FTPVPKDDLY NNLIVNPRSV GLANQELAEV VSRAVSDGYS CVTLGGDHSL AIGTISGHAR HCPDLCVVWV DAHADINTPL TTSSGNLHGQ PVSFLLRELQ DKVPQLPGFS WIKPCISSAS IVYIGLRDVD PPEHFILKNY DIQYFSMRDI DRLGIQKVME RTFDLLIGKR QRPIHLSFDI DAFDPTLAPA TGTPVVGGLT YREGMYIAEE IHNTGLLSAL DLVEVNPQLA TSEEEAKTTA NLAVDVIASS FGQTREGGHI VYDQLPTPSS PDESENQARV RI
Greater than 80% as determined by SDS-PAGE.
384
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers