missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL3 Protein
A cDNA sequence encoding the ARL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11071-1mg
Additional Details : Weight : 0.01000kg
Specifications
ARL3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGLLSI LRKLKSAPDQ EVRILLLGLD NAGKTTLLKQ LASEDISHIT PTQGFNIKSV QSQGFKLNVW DIGGQRKIRP YWKNYFENTD ILIYVIDSAD RKRFEETGQE LAELLEEEKL SCVPVLIFAN KQDLLTAAPA SEIAEGLNLH TIRDRVWQIQ SCSALTGEGV QDGMNWVCKN VNAKKK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ARL3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.1M NaCl, 1mM DTT and 0.1mM PMSF. |