missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASS1 Protein
A cDNA sequence encoding the ASS1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | ASS1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15996118
|
enQuireBio™
QP11094-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976118
|
enQuireBio™
QP11094-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986118
|
enQuireBio™
QP11094-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ASS1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ASS1 Human 0.5 mg/ml solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl, 1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSKGSVVLA YSGGLDTSCI LVWLKEQGYD VIAYLANIGQ KEDFEEARKK ALKLGAKKVF IEDVSREFVE EFIWPAIQSS ALYEDRYLLG TSLARPCIAR KQVEIAQREG AKYVSHGATG KGNDQVRFEL SCYSLAPQIK VIAPWRMPEF YNRFKGRNDL MEYAKQHGIP IPVTPKNPWS MDENLMHISY EAGILENPKN QAPPGLYTKT QDPAKAPNTP DILEIEFKKG VPVKVTNVKD GTTHQTSLEL FMYLNEVAGK HGVGRIDIVE NRFIGMKSRG IYETPAGTIL YHAHLDIEAF TMDREVRKIK QGLGLKFAEL VYTGFWHSPE CEFVRHCIAK SQERVEGKVQ VSVLKGQVYI LGRESPLSLY NEELVSMNVQ GDYEPTDATG FININSLRLK EYHRLQSKVT AK | |
Greater than 90.0% as determined by SDS-PAGE. |