missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATF2 Protein
A DNA sequence encoding the Homo sapiens (Human) ATF2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001243019.1 |
---|---|
Gene ID (Entrez) | 1386 |
Name | ATF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927019
|
enQuireBio™
QP5688-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947019
|
enQuireBio™
QP5688-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937019
|
enQuireBio™
QP5688-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957019
|
enQuireBio™
QP5688-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967019
|
enQuireBio™
QP5688-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977019
|
enQuireBio™
QP5688-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001243019.1 | |
ATF2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1386 | |
Research Use Only | |
ATF2 | |
Recombinant Protein | |
E. coli | |
MKFKLHVNSA RQYKDLWNMS DDKPFLCTAP GCGQRFTNED HLAVHKHKHE MTLKFGPARN DSVIVADQTP TPTRFLKNCE EVGLFNELAS PFENEFKKAS EDDIKKMPLD LSPLATPIIR SKIEEPSVVE TTHQDSPLPH PESTTSDEKE VPLAQTAQPT SAIVRPASLQ VPNVLLTSSD SSVIIQQAVP SPTSSTVITQ APSSNRPIVP VPGPFPLLLH LPNGQTMPVA IPASITSSNV HVPAAVPLVR PVTMVPSVPG IPGPSSPQPV QSEAKMRLKA ALTQQHPPVT NGDTVKGHGS GLVRTQSEES RPQSLQQPAT STTETPASPA HTTPQTQSTS GRRRRAANED PDEKRRKFLE RNRAAASRCR QKRKVWVQSL EKKAEDLSSL NGQLQSEVTL LRNEVAQLKQ LLLAHKDCPV TAMQKKSGYH TADKDDSSED ISVPSSPHTE AIQHSSVSTS NGVSSTSKAE AVATSVLTQM ADQSTEPALS QIVMAPSSQS QPSGS | |
Greater than 90% as determined by SDS-PAGE. |