missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP6V1F Protein
A DNA sequence encoding the Homo sapiens (Human) ATP6V1F, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP909-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001185838.1 | |
ATP6V1F Protein | |
Research Use Only | |
ATP6V1F | |
Recombinant Protein | |
E. coli | |
MAGRGKLIAV IGDEDTVTGF LLGGIGELNK NRHPNFLVVE KDTTINEIED TFRQFLNRDD IGIILINQYI AEMVRHALDA HQQSIPAVLE IPSKEHPYDA AKDSILRRAR GMFTAEDLR | |
Greater than 90% as determined by SDS-PAGE. |
9296 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |