missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAG family molecular chaperone regulator 2 Protein
A DNA sequence encoding the Homo sapiens (Human) BAG family molecular chaperone regulator 2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_004273.1 |
---|---|
Gene ID (Entrez) | 9532 |
Name | BAG family molecular chaperone regulator 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907129
|
enQuireBio™
QP5703-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927129
|
enQuireBio™
QP5703-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917129
|
enQuireBio™
QP5703-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15937129
|
enQuireBio™
QP5703-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15947129
|
enQuireBio™
QP5703-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957129
|
enQuireBio™
QP5703-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004273.1 | |
BAG family molecular chaperone regulator 2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9532 | |
Research Use Only | |
BAG2 | |
Recombinant Protein | |
E. coli | |
AQAKINAKAN EGRFCRSSSM ADRSSRLLES LDQLELRVEA LREAATAVEQ EKEILLEMIH SIQNSQDMRQ ISDGEREELN LTANRLMGRT LTVEVSVETI RNPQQQESLK HATRIIDEVV NKFLDDLGNA KSHLMSLYSA CSSEVPHGPV DQKFQSIVIG CALEDQKKIK RRLETLLRNI ENSDKAIKLL EHSKGAGSKT LQQNAESRFN | |
Greater than 80% as determined by SDS-PAGE. |