missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCHE / Butyrylcholinesterase Protein

Product Code. p-7155578
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16087431

Brand: enQuireBio™ QP9007ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) BCHE / Butyrylcholinesterase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000046.1
Gene ID (Entrez) 590
Name BCHE / Butyrylcholinesterase Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol BCHE
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence EDDIIIATKN GKVRGMNLTV FGGTVTAFLG IPYAQPPLGR LRFKKPQSLT KWSDIWNATK YANSCCQNID QSFPGFHGSE MWNPNTDLSE DCLYLNVWIP APKPKNATVL IWIYGGGFQT GTSSLHVYDG KFLARVERVI VVSMNYRVGA LGFLALPGNP EAPGNMGLFD QQLALQWVQK NIAAFGGNPK SVTLFGESAG AASVSLHLLS PGSHSLFTRA ILQSGSFNAP WAVTSLYEAR NRTLNLAKLT GCSRENETEI IKCLRNKDPQ EILLNEAFVV PYGTPLSVNF GPTVDGDFLT DMPDILLELG QFKKTQILVG VNKDEGTAFL VYGAPGFSKD NNSIITRKEF QEGLKIFFPG VSEFGKESIL FHYTDWVDDQ RPENYREALG DVVGDYNFIC PALEFTKKFS EWGNNAFFYY FEHRSSKLPW PEWMGVMHGY EIEFVFGLPL ERRDNYTKAE EILSRSIVKR WANFAKYGNP NETQNNSTSW PVFKSTEQKY LTLNTESTRI MTKLRAQQCR FWTSFFPKVL EMTGNIDEAE WEWKAGFHRW NNYMMDWKNQ FNDYTSKKES CVGL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.