missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Bcl-2 related protein A1 (aa 1-152)
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Bcl-2 related protein A1 (aa 1-152) is derived from E. coli. The Recombinant Human Bcl-2 related protein A1 (aa 1-152) has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol. |
| Gene ID (Entrez) | 597 |
| Molecular Weight (g/mol) | 18 kDa |
| Name | Bcl-2 related protein A1 Minus C-terminus, aa 1-152 |
| Quantity | 50 μg |
| Source | E. coli-derived human Bcl-2 related protein A1 protein Thr2-Ser152,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction