missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Bcl-w (minus C-Terminus) Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Bcl-w (minus C-Terminus) Protein is derived from E. coli. The Recombinant Human Bcl-w (minus C-Terminus) Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl and Glycerol. |
| Gene ID (Entrez) | 599 |
| Molecular Weight (g/mol) | 19.5 kDa |
| Name | Bcl-w Minus C-Terminus |
| Quantity | 50 μg |
| Source | E. coli-derived human Bcl-w protein Ala2-Thr172,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Monomer and disulfide-linked homodimer |
| Endotoxin Concentration | <0.01 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction