missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-1,4-galactosyltransferase 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Beta-1,4-galactosyltransferase 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8332-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16014061

  • 505.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001186802.1
Beta-1,4-galactosyltransferase 3 Protein
Research Use Only
B4GALT3
Recombinant Protein
E. coli
LSALFGRDQG PTFDYSHPRD VYSNLSHLPG APGGPPAPQG LPYCPERSPL LVGPVSVSFS PVPSLAEIVE RNPRVEPGGR YRPAGCEPRS RTAIIVPHRA REHHLRLLLY HLHPFLQRQQ LAYGIYVIHQ AGNGTFNRAK LLNVGVREAL RDEEWDCLFL HDVDLLPEND HNLYVCDPRG PRHVAVAMNK FGYSLPYPQY FGGVSALTPD QYLKMNGFPN EYWGWGGEDD DIATRVRLAG MKISRPPTSV GHYKMVKHRG DKGNEENPHR FDLLVRTQNS WTQDGMNSLT YQLLARELGP LYTNITADIG TDPRGPRAPS GPRYPPGSSQ AFRQEMLQRR PPARPGPLST ANHTALRGSH
Greater than 90% as determined by SDS-PAGE.
8703
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers