missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BHMT Protein
A cDNA sequence encoding the BHMT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11159-50ug
Additional Details : Weight : 0.01000kg
Specifications
BHMT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMPP VGGKKAKKGI LERLNAGEIV IGDGGFVFAL EKRGYVKAGP WTPEAAVEHP EAVRQLHREF LRAGSNVMQT FTFYASEDKL ENRGNYVLEK ISGQEVNEAA CDIARQVADE GDALVAGGVS QTPSYLSCKS ETEVKKVFLQ QLEVFMKKNV DFLIAEYFEH VEEAVWAVET LIASGKPVAA TMCIGPEGDL HGVPPGECAV RLVKAGASII GVNCHFDPTI SLKTVKLMKE GLEAARLKAH LMSQPLAYHT PDCNKQGFIDLPEFPFGLEP RVATRWDIQK YAREAYNLGV RYIGGCCGFE PYHIRAIAEE LAPERGFLPPASEKHGSWGS GLDMHTKPWV RARARKEYWE NLRIASGRPY NPSMSKPDGW GVTKGTAELM QQKEATTEQQ LKELFEKQKF KSQ | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The BHMT solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol. |