missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bifunctional epoxide hydrolase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Bifunctional epoxide hydrolase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5986-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243411.1 | |
Bifunctional epoxide hydrolase 2 Protein | |
Research Use Only | |
EPHX2 | |
Recombinant Protein | |
E. coli | |
MTLRAAVFDL DGVLALPAVF GVLGRTEEAL ALPRGLLNDA FQKGGPEGAT TRLMKGEITL SQWIPLMEEN CRKCSETAKV CLPKNFSIKE IFDKAISARK INRPMLQAAL MLRKKGFTTA ILTNTWLDDR AERDGLAQLM CELKMHFDFL IESCQVGMVK PEPQIYKFLL DTLKASPSEV VFLDDIGANL KPARDLGMVT ILVQDTDTAL KELEKVTGIQ LLNTPAPLPT SCNPSDMSHG YVTVKPRVRL HFVELGSGPA VCLCHGFPES WYSWRYQIPA LAQAGYRVLA MDMKGYGESS APPEIEEYCM EVLCKEMVTF LDKLGLSQAV FIGHDWGGML VWYMALFYPE RVRAVASLNT PFIPANPNMS PLESIKANPV FDYQLYFQEP GVAEAELEQN LSRTFKSLFR ASDESVLSMH KVCEAGGLFV NSPEEPSLSR MVTEEEIQFY VQQFKKSGFR GPLNWYRNME RNWKWACKSL GRKILIPALM VTAEKDFVLV PQMSQHMEDW IPHLKRGHIE DCGHWTQMDK PTEVNQILIK WLDSDARNPP VVSKM | |
Greater than 90% as determined by SDS-PAGE. |
2053 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |