missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BMP-4 (E. coli-expressed) Protein

Product Code. 18786466
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18786466 50 μg 50µg
18775746 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18786466 Supplier R&D Systems™ Supplier No. 314BPE050
  • ONLINE EXCLUSIVE PRICE
    721.29 €
    647.85 € / 50µg
    Save 73.44 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12644
For Use With (Application) Bioactivity
Gene ID (Entrez) 652
Molecular Weight (g/mol) 12 kDa
Name BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Quantity 50 μg
Source E. coli-derived human BMP-4 protein Lys303-Arg408
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.