missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMPR-IA/ALK-3 Fc Chimera Protein
missing translation for 'orderingAttributeHoverText'
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
missing translation for 'unitSize'
100µg
Beschreibung
The Recombinant Human BMPR-IA/ALK-3 Fc Chimera Protein is derived from NS0. The Recombinant Human BMPR-IA/ALK-3 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 657 |
| Molecular Weight (g/mol) | 41.5 kDa (monomer) |
| Name | BMPR-IA/ALK-3 Fc Chimera |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line,NS0-derived human BMPR-IA/ALK-3 protein, (N-terminus) Human BMPR-IA (Gln24-Arg152) Accession &Num; CAA80257 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur