missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BNIP3L Protein
A DNA sequence encoding the Homo sapiens (Human) BNIP3L, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001317420.1 |
---|---|
Gene ID (Entrez) | 665 |
Name | BNIP3L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037441
|
enQuireBio™
QP9010-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 10-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077441
|
enQuireBio™
QP9010-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047441
|
enQuireBio™
QP9010-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067441
|
enQuireBio™
QP9010-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087441
|
enQuireBio™
QP9010-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057441
|
enQuireBio™
QP9010-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001317420.1 | |
BNIP3L Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
665 | |
Research Use Only | |
BNIP3L | |
Recombinant Protein | |
Yeast | |
MSSHLVEPPP PLHNNNNNCE ENEQSLPPPA GLNSSWVELP MNSSNGNDNG NGKNGGLEHV PSSSSIHNGD MEKILLDAQH ESGQSSSRGS SHCDSPSPQE DGQIMFDVEM HTSRDHSSQS EEEVVEGEKE VEALKKSADW VSDWSSRPEN IPPKEFHFRH PKRSVSLSMR KSGAMKKGGI FSAEFLKVFI PSLFLSHVLA LGLGIYIGKR LSTPSASTY | |
Greater than 90% as determined by SDS-PAGE. |