missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRAK Protein
A cDNA sequence encoding the BRAK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11201-50ug
Additional Details : Weight : 0.01000kg
Specifications
9547 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIITTKS VSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
BRAK Protein | |
Research Use Only | |
CXCL14 | |
Human | |
His | |
CXCL14 filtered (0.4microm) and lyophilized from a concentrated (0.5 mg/ml) solution containing 20mM Tris buffer and 20mM NaCl pH 7.5. |