missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Breast carcinoma-amplified sequence 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Breast carcinoma-amplified sequence 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5705-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003648.2 | |
Breast carcinoma-amplified sequence 1 Protein | |
Research Use Only | |
BCAS1 | |
Recombinant Protein | |
E. coli | |
MGNQMSVPQR VEDQENEPEA ETYQDNASAL NGVPVVVSTH TVQHLEEVDL GISVKTDNVA TSSPETTEIS AVADANGKNL GKEAKPEAPA AKSRFFLMLS RPVPGRTGDQ AADSSLGSVK LDVSSNKAPA NKDPSESWTL PVAAGPGQDT DKTPGHAPAQ DKVLSAARDP TLLPPETGGA GGEAPSKPKD SSFFDKFFKL DKGQEKVPGD SQQEAKRAEH QDKVDEVPGL SGQSDDVPAG KDIVDGKEKE GQELGTADCS VPGDPEGLET AKDDSQAAAI AENNNSIMSF FKTLVSPNKA ETKKDPEDTG AEKSPTTSAD LKSDKANFTS QETQGAGKNS KGCNPSGHTQ SVTTPEPAKE GTKEKSGPTS LPLGKLFWKK SVKEDSVPTG AEENVVCESP VEIIKSKEVE SALQTVDLNE GDAAPEPTEA KLKREESKPR TSLMAFLRQM SVKGDGGITH SEEINGKDSS CQTSDSTEKT ITPPEPEPTG APQKGKEGSS KDKKSAAEMN KQKSNKQEAK EPAQCTEQAT VDTNSLQNGD KLQKRPEKRQ QSLGGFFKGL GPKRMLDAQV QTDPVSIGPV GKSK | |
Greater than 90% as determined by SDS-PAGE. |
8537 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |