missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BST2 Protein
A cDNA sequence encoding the BST2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11208-10ug
This item is not returnable.
View return policy
Specifications
684 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSEACRDGLR AVMECRNVTH LLQQELTEAQ KGFQDVEAQA ATCNHTVMAL MASLDAEKAQ GQKKVEELEG EITTLNHKLQ DASAEVERLR RENQVLSVRI ADKKYYPSSQ DSS | |
Greater than 80.0% as determined by SDS-PAGE. |
BST2 Protein | |
Research Use Only | |
BST2 | |
Human | |
His | |
BST2 protein 0.5 mg/ml is supplied in 20mM Tris-HCl, pH 8.0, 0.1M NaCl, 1mM DTT and 20% Glycerol. |