missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Butyrophilin subfamily 2 member A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Butyrophilin subfamily 2 member A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7953-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001184166.1 | |
Butyrophilin subfamily 2 member A2 Protein | |
Research Use Only | |
BTN2A2 | |
Recombinant Protein | |
E. coli | |
QFTVVGPANP ILAMVGENTT LRCHLSPEKN AEDMEVRWFR SQFSPAVFVY KGGRERTEEQ MEEYRGRITF VSKDINRGSV ALVIHNVTAQ ENGIYRCYFQ EGRSYDEAIL RLVVAGLGSK PLIEIKAQED GSIWLECISG GWYPEPLTVW RDPYGEVVPA LKEVSIADAD GLFMVTTAVI IRDKYVRNVS CSVNNTLLGQ EKETVIFIPE SFMPSASPWM VALAVILTAS | |
Greater than 90% as determined by SDS-PAGE. |
10385 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |