missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-JUN Protein
A cDNA sequence encoding the C-JUN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10551-10ug
Additional Details : Weight : 0.01000kg
Specifications
3725 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
C-Jun is phosphorylatable in vitro, using either recombinant active JNK1 or JNK2, or with JNK immunoprecipitated from stimulated cells. This phosphorylation can be monitored by Western blot analysis using an antibody directed to c-Jun [pS73], in conjunction with chemiluminescence detection methods. Optimization of the cell stimulation protocol, cell lysis procedure, and reaction conditions may be required for each specific application. | |
Human | |
His | |
C-JUN is supplied as lyophilized powder containing no additives. |
C-JUN Protein | |
Research Use Only | |
JUN | |
Recombinant Protein | |
E. coli | |
MTAKMETTFY DDALNASFLP SESGPYGYSN PKILKQSMTL NLADPVGSLK PHLRAKNSDL LTSPDVGLLK LASPELERLI IQSSNGHITT TPTPTQFLCP KNVTDEQEGF AEGFVRALAE LHSQNTLPSV TSAAQPVNGA GMVAPAVASV AGGSGSGGFS ASLHSEPPVY ANLSNFNPGA LSSGGGAPSY GAAGLAFPAQ PQQQQQPPHH LPQQMPVQHP RLQALKEEPQ TVPEMPGETP PLSPIDMESQ ERIKAERKRM RNRIAASKCR KRKLERIARL EEKVKTLKAQ NSELASTANM LREQVAQLKQ KVMNHVNSGC QLMLTQQLQT F | |
Greater than 95% as determined by SDS-PAGE. |