missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-JUN Protein

A cDNA sequence encoding the C-JUN was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10551-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15994218

  • 131.00 € / 2µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3725
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
C-Jun is phosphorylatable in vitro, using either recombinant active JNK1 or JNK2, or with JNK immunoprecipitated from stimulated cells. This phosphorylation can be monitored by Western blot analysis using an antibody directed to c-Jun [pS73], in conjunction with chemiluminescence detection methods. Optimization of the cell stimulation protocol, cell lysis procedure, and reaction conditions may be required for each specific application.
Human
His
C-JUN is supplied as lyophilized powder containing no additives.
C-JUN Protein
Research Use Only
JUN
Recombinant Protein
E. coli
MTAKMETTFY DDALNASFLP SESGPYGYSN PKILKQSMTL NLADPVGSLK PHLRAKNSDL LTSPDVGLLK LASPELERLI IQSSNGHITT TPTPTQFLCP KNVTDEQEGF AEGFVRALAE LHSQNTLPSV TSAAQPVNGA GMVAPAVASV AGGSGSGGFS ASLHSEPPVY ANLSNFNPGA LSSGGGAPSY GAAGLAFPAQ PQQQQQPPHH LPQQMPVQHP RLQALKEEPQ TVPEMPGETP PLSPIDMESQ ERIKAERKRM RNRIAASKCR KRKLERIARL EEKVKTLKAQ NSELASTANM LREQVAQLKQ KVMNHVNSGC QLMLTQQLQT F
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers