missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-Myc-binding Protein

A DNA sequence encoding the Homo sapiens (Human) C-Myc-binding protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8016-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042171

  • 505.00 € / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036465.2
C-Myc-binding Protein
Research Use Only
MYCBP
Recombinant Protein
E. coli
AHYKAADSKR EQFRRYLEKS GVLDTLTKVL VALYEEPEKP NSALDFLKHH LGAATPENPE IELLRLELAE MKEKYEAIVE ENKKLKAKLA QYEPPQEEKR AE
Greater than 90% as determined by SDS-PAGE.
26292
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers