missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-X-C chemokine receptor type 2 Protein
A DNA sequence encoding the Homo sapiens (Human) C-X-C chemokine receptor type 2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001161770.1 |
---|---|
Gene ID (Entrez) | 3579 |
Name | C-X-C chemokine receptor type 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031770
|
enQuireBio™
QP6229-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051770
|
enQuireBio™
QP6229-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041770
|
enQuireBio™
QP6229-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061770
|
enQuireBio™
QP6229-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071770
|
enQuireBio™
QP6229-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081770
|
enQuireBio™
QP6229-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001161770.1 | |
C-X-C chemokine receptor type 2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3579 | |
Research Use Only | |
CXCR2 | |
Recombinant Protein | |
E. coli | |
MEDFNMESDS FEDFWKGEDL SNYSYSSTLP PFLLDAAPCE | |
Greater than 90% as determined by SDS-PAGE. |