missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF9 Protein
A cDNA sequence encoding the C1QTNF9 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifica
Name | C1QTNF9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
15906578
|
enQuireBio™
QP11236-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986568
|
enQuireBio™
QP11236-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996568
|
enQuireBio™
QP11236-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifica
C1QTNF9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C1QTNF9 Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 0.03M Acetate buffer, pH 4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS QDTCRQGHPG IPGNPGHNGL PGRDGRDGAK GDKGDAGEPG RPGSPGKDGT SGEKGERGAD GKVEAKGIKG DQGSRGSPGK HGPKGLAGPM GEKGLRGETG PQGQKGNKGD VGPTGPEGPR GNIGPLGPTG LPGPMGPIGK PGPKGEAGPT GPQGEPGVRG IRGWKGDRGE KGKIGETLVL PKSAFTVGLT VLSKFPSSDM PIKFDKILYN EFNHYDTAAG KFTCHIAGVY YFTYHITVFS RNVQVSLVKN GVKILHTKDA YMSSEDQASG GIVLQLKLGD EVWLQVTGGE RFNGLFADED DDTTFTGFLL FSSP |