missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C4BPB Protein
A cDNA sequence encoding the C4BPB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11239-1mg
Additional Details : Weight : 0.01000kg
Specifications
C4BPB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSDAEH CPELPPVDNS IFVAKEVEGQ ILGTYVCIKG YHLVGKKTLF CNASKEWDNT TTECRLGHCP DPVLVNGEFS SSGPVNVSDK ITFMCNDHYI LKGSNRSQCL EDHTWAPPFP ICKSRDCDPP GNPVHGYFEG NNFTLGSTIS YYCEDRYYLV GVQEQQCVDG EWSSALPVCK LIQEAPKPEC EKALLAFQES KNLCEAMENF MQQLKESGMT MEELKYSLEL KKAELKAKLL | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C4BPB protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol and 0.15M NaCl. |