missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C4BPB Protein

A cDNA sequence encoding the C4BPB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11239-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986578

  • 203.00 € / 25µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C4BPB Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSDAEH CPELPPVDNS IFVAKEVEGQ ILGTYVCIKG YHLVGKKTLF CNASKEWDNT TTECRLGHCP DPVLVNGEFS SSGPVNVSDK ITFMCNDHYI LKGSNRSQCL EDHTWAPPFP ICKSRDCDPP GNPVHGYFEG NNFTLGSTIS YYCEDRYYLV GVQEQQCVDG EWSSALPVCK LIQEAPKPEC EKALLAFQES KNLCEAMENF MQQLKESGMT MEELKYSLEL KKAELKAKLL
Greater than 90% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
C4BPB protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol and 0.15M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers