missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CA3 Protein

A cDNA sequence encoding the CA3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11248-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956608

  • 203.00 € / 20µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

761
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MAKEWGYASH NGPDHWHELF PNAKGENQSP IELHTKDIRH DPSLQPWSVS YDGGSAKTIL NNGKTCRVVF DDTYDRSMLR GGPLPGPYRL RQFHLHWGSS DDHGSEHTVD GVKYAAELHL VHWNPKYNTF KEALKQRDGI AVIGIFLKIG HENGEFQIFL DALDKIKTKG KEAPFTKFDP SCLFPACRDY WTYQGSFTTP PCEECIVWLL LKEPMTVSSD QMAKLRSLLS SAENEPPVPL VSNWRPPQPI NNRVVRASFK
Greater than 90% as determined by SDS-PAGE.
CA3 Protein
Research Use Only
CA3
Human
Untagged
The CA3 solution contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers