missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAIII Protein
A cDNA sequence encoding the CAIII was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Gene ID (Entrez) | 761 |
---|---|
Name | CAIII Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CA3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906638
|
enQuireBio™
QP11256-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996628
|
enQuireBio™
QP11256-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986628
|
enQuireBio™
QP11256-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
761 | |
Research Use Only | |
CA3 | |
Human | |
His | |
Supplied in 10mM Tris-HCl (pH 8), 250mM NaCl, 0.5mM DTT, 1.5mM Cysteine, and 50% Glycerol. |
CAIII Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAKEWGYASH NGPDHWHELF PNAKGENQSP VELHTKDIRH DPSLQPWSVS YDGGSAKTIL NNGKTCRVVF DDTYDRSMLR GGPLPGPYRL RQFHLHWGSS DDHGSEHTVD GVKYAAELHL VHWNPKYNTF KEALKQRDGI AVIGIFLKIG HENGEFQIFL DALDKIKTKG KEAPFTKFDP SCLFPACRDY WTYQGSFTTP PCEECIVWLL LKEPMTVSSD QMAKLRSLLS SAENEPPVPL VSNWRPPQPI NNRVVRASFK | |
Greater than 95.0% as determined by SDS-PAGE. |