missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALCA / CGRP Protein
Recombinant Protein
107.00 € - 7105.00 €
Specifications
Accession Number | NP_001029124.1 |
---|---|
Gene ID (Entrez) | 796 |
Name | CALCA / CGRP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985849
|
enQuireBio™
QP5287-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965849
|
enQuireBio™
QP5287-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955849
|
enQuireBio™
QP5287-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995849
|
enQuireBio™
QP5287-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975849
|
enQuireBio™
QP5287-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001029124.1 | |
CALCA / CGRP Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APFRSALESS PADPATLSED EARLLLAALV QDYVQMKASE LEQEQEREGS SLDSPRSKRC GNLSTCMLGT YTQDFNKFHT FPQTAIGVGA PGKKRDMSSD LERDHRPHVS MPQNAN | |
Greater than 95% as determined by SDS-PAGE. |
796 | |
Research Use Only | |
CALCA | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |