missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Calcium-dependent phospholipase A2 Protein

A DNA sequence encoding the Homo sapiens (Human) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9333-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038111

  • 1665.00 € / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000920.1
Calcium-dependent phospholipase A2 Protein
Research Use Only
PLA2G5
Recombinant Protein
Yeast
GLLDLKSMIE KVTGKNALTN YGFYGCYCGW GGRGTPKDGT DWCCWAHDHC YGRLEEKGCN IRTQSYKYRF AWGVVTCEPG PFCHVNLCAC DRKLVYCLKR NLRSYNPQYQ YFPNILCS
Greater than 90% as determined by SDS-PAGE.
5322
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers