missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Calmodulin Protein

A DNA sequence encoding the Homo sapiens (Human) Calmodulin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9025-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037481

  • 1730.00 € / 500µg
Estimated Shipment: 04-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001292553.1
Calmodulin Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
801
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CALM1
Recombinant Protein
Yeast
ADQLTEEQIA EFKEAFSLFD KDGDGTITTK ELGTVMRSLG QNPTEAELQD MINEVDADGN GTIDFPEFLT MMARKMKDTD SEEEIREAFR VFDKDGNGYI SAAELRHVMT NLGEKLTDEE VDEMIREADI DGDGQVNYEE FVQMMTAK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers