missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpain-14 Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain-14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10031-ba-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138594.1 | |
Calpain-14 Protein | |
Research Use Only | |
CAPN14 | |
Recombinant Protein | |
Baculovirus | |
LFEDTSFPAT LSSIGSGSLL QKLPPRLQWK RPPELHSNPQ FYFAKAKRLD LCQGIVGDCW FLAALQALAL HQDILSRVVP LNQSFTEKYA GIFRFWFWHY GNWVPVVIDD RLPVNEAGQL VFVSSTYKNL FWGALLEKAY AKLSGSYEDL QSGQVSEALV DFTGGVTMTI NLAEAHGNLW DILIEATYNR TLIGCQTHSG EKILENGLVE GHAYTLTGIR KVTCKHRPEY LVKLRNPWGK VEWKGDWSDS SSKWELLSPK EKILLLRKDN DGEFWMTLQD FKTHFVLLVI CKLTPGLLSQ EAAQKWTYTM REGRWEKRST AGGQRQLLQD TFWKNPQFLL SVWRPEEGRR SLRPCSVLVS LLQKPRHRCR KRKPLLAIGF YLYRMNKYHD DQRRLPPEFF QRNTPLSQPD RFLKEKEVSQ ELCLEPGTYL IVPCILEAHQ KSEFVLRVFS RKHIFYEIGS NSGVVFSKEI EDQNERQDEF FTKFFEKHPE INAVQLQNLL NQMTWSSLGS RQPFFSLEAC QGILALLDLN ASGTMSIQEF RDLWKQLKLS QKVFHKQDRG SGYLNWEQLH AAMREAGIML SDDVCQLMLI RYGGPRLQMD FVSFIHLMLR VENMEDVFQN LTQDGKGIYL QKPEWMMMAL YS | |
Greater than 80% as determined by SDS-PAGE. |
440854 | |
20 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |