missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calponin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Calponin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5848-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001290.2 | |
Calponin-1 Protein | |
Research Use Only | |
CNN1 | |
Recombinant Protein | |
E. coli | |
MSSAHFNRGP AYGLSAEVKN KLAQKYDHQR EQELREWIEG VTGRRIGNNF MDGLKDGIIL CEFINKLQPG SVKKINESTQ NWHQLENIGN FIKAITKYGV KPHDIFEAND LFENTNHTQV QSTLLALASM AKTKGNKVNV GVKYAEKQER KFEPGKLREG RNIIGLQMGT NKFASQQGMT AYGTRRHLYD PKLGTDQPLD QATISLQMGT NKGASQAGMT APGTKRQIFE PGLGMEHCDT LNVSLQMGSN KGASQRGMTV YGLPRQVYDP KYCLTPEYPE LGEPAHNHHA HNYYNSA | |
Greater than 90% as determined by SDS-PAGE. |
1264 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |