missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCR4-NOT transcription complex subunit 8 Protein

Product Code. 16042681
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16042681 100 μg 100µg
16052681 10 μg 10µg
16092681 50 μg 50µg
16072681 200 μg 200µg
16082681 500 μg 500µg
16062681 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16042681 Supplier enQuireBio™ Supplier No. QP8105ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) CCR4-NOT transcription complex subunit 8, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001288002.1
Gene ID (Entrez) 9337
Name CCR4-NOT transcription complex subunit 8 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CNOT8
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MPAALVENSQ VICEVWASNL EEEMRKIREI VLSYSYIAMD TEFPGVVVRP IGEFRSSIDY QYQLLRCNVD LLKIIQLGLT FTNEKGEYPS GINTWQFNFK FNLTEDMYSQ DSIDLLANSG LQFQKHEEEG IDTLHFAELL MTSGVVLCDN VKWLSFHSGY DFGYMVKLLT DSRLPEEEHE FFHILNLFFP SIYDVKYLMK SCKNLKGGLQ EVADQLDLQR IGRQHQAGSD SLLTGMAFFR MKELFFEDSI DDAKYCGRLY GLGTGVAQKQ NEDVDSAQEK MSILAIINNM QQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.