missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD10 / Neprilysin / MME Protein
A DNA sequence encoding the Homo sapiens (Human) CD10 / Neprilysin / MME, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_000893.2 |
---|---|
Gene ID (Entrez) | 4311 |
Name | CD10 / Neprilysin / MME Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097981
|
enQuireBio™
QP9276-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037991
|
enQuireBio™
QP9276-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007991
|
enQuireBio™
QP9276-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027991
|
enQuireBio™
QP9276-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047991
|
enQuireBio™
QP9276-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017991
|
enQuireBio™
QP9276-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000893.2 | |
CD10 / Neprilysin / MME Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4311 | |
Research Use Only | |
MME | |
Recombinant Protein | |
Yeast | |
YDDGICKSSD CIKSAARLIQ NMDATTEPCT DFFKYACGGW LKRNVIPETS SRYGNFDILR DELEVVLKDV LQEPKTEDIV AVQKAKALYR SCINESAIDS RGGEPLLKLL PDIYGWPVAT ENWEQKYGAS WTAEKAIAQL NSKYGKKVLI NLFVGTDDKN SVNHVIHIDQ PRLGLPSRDY YECTGIYKEA CTAYVDFMIS VARLIRQEER LPIDENQLAL EMNKVMELEK EIANATAKPE DRNDPMLLYN KMTLAQIQNN FSLEINGKPF SWLNFTNEIM STVNISITNE EDVVVYAPEY LTKLKPILTK YSARDLQNLM SWRFIMDLVS SLSRTYKESR NAFRKALYGT TSETATWRRC ANYVNGNMEN AVGRLYVEAA FAGESKHVVE DLIAQIREVF IQTLDDLTWM DAETKKRAEE KALAIKERIG YPDDIVSNDN KLNNEYLELN YKEDEYFENI IQNLKFSQSK QLKKLREKVD KDEWISGAAV VNAFYSSGRN QIVFPAGILQ PPFFSAQQSN SLNYGGIGMV IGHEITHGFD DNGRNFNKDG DLVDWWTQQS ASNFKEQSQC MVYQYGNFSW DLAGGQHLNG INTLGENIAD NGGLGQAYRA YQNYIKKNGE EKLLPGLDLN HKQLFFLNFA QVWCGTYRPE YAVNSIKTDV HSPGNFRIIG TLQNSAEFSE AFHCRKNSYM NPEKKCRVW | |
Greater than 90% as determined by SDS-PAGE. |