missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD207 Protein
A cDNA sequence encoding the CD207 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11314-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
50489 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSPRFMGTI SDVKTNVQLL KGRVDNISTL DSEIKKNSDG MEAAGVQIQM VNESLGYVRS QFLKLKTSVE KANAQIQILT RSWEEVSTLN AQIPELKSDL EKASALNTKI RALQGSLENM SKLLKRQNDI LQVVSQGWKY FKGNFYYFSL IPKTWYSAEQ FCVSRNSHLT SVTSESEQEF LYKTAGGLIY WIGLTKAGME GDWSWVDDTP FNKVQSARFW IPGEPNNAGN NEHCGNIKAP SLQAWNDAPC DKTFLFICKR PYVPSEP | |
Greater than 85.0% as determined by SDS-PAGE. |
CD207 Protein | |
Research Use Only | |
CD207 | |
Human | |
His | |
CD207 protein solution (1 mg/ml) containing 20mM Tris 8.0 and 10% glycerol. |