missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD244 Protein

A cDNA sequence encoding the CD244 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11316-HIS-INSECT-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956828

  • 131.00 € / 2µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

51744
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
MGSSHHHHHH SSGLVPRGSH MGSHGKGCQG SADHVVSISG VPLQLQPNSI QTKVDSIAWK KLLPSQNGFH HILKWENGSL PSNTSNDRFS FIVKNLSLLI KAAQQQDSGL YCLEVTSISG KVQTATFQVF VFDKVEKPRL QGQGKILDRG RCQVALSCLV SRDGNVSYAW YRGSKLIQTA GNLTYLDEEV DINGTHTYTC NVSNPVSWES HTLNLTQDCQ NAHQEFRFWP
Greater than 85% as determined by SDS-PAGE.
CD244 Protein
Research Use Only
CD244
Human
His
CD244 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0 and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers