missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD274 Protein
A cDNA sequence encoding the CD274 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 29126 |
---|---|
Name | CD274 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD274 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976838
|
enQuireBio™
QP11319-HIS-EC-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966838
|
enQuireBio™
QP11319-HIS-EC-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956838
|
enQuireBio™
QP11319-HIS-EC-100UG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
29126 | |
Research Use Only | |
CD274 | |
Human | |
His | |
The CD274 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |
CD274 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMFTVTV PKDLYVVEYG SNMTIECKFP VEKQLDLAAL IVYWEMEDKN IIQFVHGEED LKVQHSSYRQ RARLLKDQLS LGNAALQITD VKLQDAGVYR CMISYGGADY KRITVKVNAP YNKINQRILV VDPVTSEHEL TCQAEGYPKA EVIWTSSDHQ VLSGKTTTTN SKREEKLFNV TSTLRINTTT NEIFYCTFRR LDPEENHTAE LVIPELPLAH PPNER | |
Greater than 90% as determined by SDS-PAGE. |